Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Tritylaniline, 97%

Catalog No. AAA1055014
Click to view available options
Quantity:
5 g
25 g

CAS: 22948-06-7 | C25H21N | 335.45 g/mol

4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22948-06-7
Molecular Formula C25H21N
Molecular Weight (g/mol) 335.45
MDL Number MFCD00007898
InChI Key XYHDHXBLSLSXSR-UHFFFAOYSA-N
Synonym p-tritylaniline, benzenamine, 4-triphenylmethyl, 4-triphenylmethyl aniline, p-aminotetraphenylmethane, 4-aminotetraphenylmethane, triphenyl p-aminophenyl methane, 4-trityl aniline, 4-triphenylmethyl benzenamine, p-toluidine, alpha,alpha,alpha-triphenyl, 4-aminophenyl triphenylmethane
PubChem CID 89914
IUPAC Name 4-tritylaniline
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)N

Specifications

Melting Point 256°C to 260°C
Quantity 25 g
Beilstein 2750328
Solubility Information Insoluble in water.
Formula Weight 335.45
Percent Purity 97%
Chemical Name or Material 4-Tritylaniline
EINECSNumber 245-346-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.