Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylaniline, 90%, technical, stabilized

Catalog No. p-3779875
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 1520-21-4 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1520-21-4
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.17
MDL Number MFCD00015329
InChI Key LBSXSAXOLABXMF-UHFFFAOYSA-N
Synonym 4-vinylaniline, 4-aminostyrene, p-aminostyrene, benzenamine, 4-ethenyl, 4-aminostryene, ccris 4326, benzenamine,4-ethenyl, p-vinylaniline, p-amino styrene, para-aminostyrene
PubChem CID 73700
IUPAC Name 4-ethenylaniline
SMILES C=CC1=CC=C(C=C1)N

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 23°C to 24°C
Color Red to Yellow
Density 1.0100g/mL
Boiling Point 249°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.6250 to 1.629
Linear Formula H2C=CHC6H4NH2
Quantity 1 g
Specific Gravity 1.01
Formula Weight 119.17
Percent Purity 90%
Grade Technical
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Vinylaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Suspected of causing cancer
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
EINECSNumber 216-185-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.