Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ Enespro™ RevoLite™ 40 cal Arc Flash Lift Front Hood
Shop All National Public Seating Corporation Products

Click to view available options
:
Size: Universal
Description
- Lift front shield makes it easy to obtain fresh air while out of the hazard zone
- Knit gussets at side provide greater range of motion and flexibility
- Premium anti-fog and scratch resistant coating on faceshield
- PureView™ faceshield technology features a greater range of visibility
- Hard hat included with slotted adapter
- Vents provide fresh air and increased auditory capabilities
Specifications
Specifications
Product Type | Arc Flash Hood |
Size | Universal |
Material | RevoLite™ |
Color | Olive Green |
Disposable | Reusable |
Certifications/Compliance | NFPA 70E, ASTM F2178, CSA Z462, OSHA 1910.269 |
Quantity | 1 Ea. |
Includes | Arc Flash Hood |
For Use With (Application) | Utilities, Manufacturing, Electrical Contractors |
Style | Hood |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction