Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 40nm Colloidal Gold Nanoparticles - 15OD
SDP

Catalog No. 2120100
Click to view available options
Quantity:
10 mL
100 mL

40nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • Narrow size distribution, consistent shape
  • Less prone to aggregation
  • Uniform shape to ensures consistent binding of antibodies

Applications: Electron microscopy visualization
Storage: Store at 4°C

Note: Kits are guaranteed for 6 months from date of receipt

Specifications

Quantity 100 mL
Product Type 40nm Colloidal Gold Nanoparticles - 15OD Antibody Labeling Kit
Content And Storage 4°C Do not freeze.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.