Learn More
Invitrogen™ BID p15 Polyclonal Antibody

Description
BID p15 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | BID p15 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 |
| Gene | BID |
| Gene Accession No. | P55957 |
| Gene Alias | 2700049M22Rik; AI875481; apoptic d; apoptic death agonist; AU022477; BH3 interacting domain death agonist; BH3-interacting domain death agonist; BH3-interacting domain death agonist p11; BH3-interacting domain death agonist p13; BH3-interacting domain death agonist p15; BID; BID isoform ES(1b); BID isoform L(2); BID isoform Si6; desmocollin type 4; FP497; HGNC:1050; Human BID coding sequence; MGC15319; MGC42355; OTTHUMP00000196197; p11 BID; p13 BID; p15 BID; p22 BID |
| Gene Symbols | BID |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.