Learn More
SHOWA™ 451 Series Natural Rubber Palm Coated Thermal-Insulated Gloves
Catalog No. 19019245 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-019-245 Large
19-019-244 Medium
19-019-246 X-Large
19-019-243 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-019-245 Supplier SHOWA™ Supplier No. 451L09
Add to Cart
Add to Cart
This item is not returnable. View return policy
Created with warmth in mind, the 451 thermal-insulated gloves are designed to be worn in cold weather.
Benifits
  • Thermal Insulation
  • Flexible
  • Breathable
  • Extra Grip
  • Seamless Knit
  • Skin-friendly
  • Abrasion-resistant
Features
  • Rough Grip
  • Ergonomic
  • Elasticated Wrist Cuff
  • Natural Rubber Palm Coating
Trades & Applications
  • Signalers
  • Brick Laying
  • Carpentry
  • Driving Machines
  • Masonry
  • Demolition
  • Shipping/Receiving
Specifications
Certifications/Compliance CE Cat II, EN 388:2016, EN 511:2006
Coated Area Palm
Construction Seamless Knit
Coating Material Natural Rubber
Color Gray
Material Cotton, Polyester
Size Large
Liner Material Acrylic
Resistant To Abrasion Resistant
Series 451