Learn More
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves
Catalog No. 19395310 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-395-310 Large
19-395-309 Medium
19-395-311 X-Large
19-395-312 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-395-310 Supplier SHOWA™ Supplier No. 477L08
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to keep hands warm and dry in the most extreme environments, the 477 cold-resistant glove combines thermal insulation with tactical grip.
Benifits
  • Seamless knit
  • Cold Resistant
  • Water-resistant
  • Flexible
  • Abrasion-resistant
  • Extra Grip
  • Thermal insulation
  • Form-fitting
  • Impermeable
Features
  • Foam grip
  • Elasticated wrist cuff
  • Ergonomic
  • Foam nitrile palm dip
  • Insulated
  • Fully-coated nitrile
Trades & Applications
  • Concreting
  • Guttering and drain pipe
  • Insulation
  • Washing and cleaning
  • Planting & repotting
  • Bottling
  • Water/Sewage treatment
  • Plumbing
  • Drainage, piping
  • Shipping/Receiving
Specifications
Certifications/Compliance CE Cat II, EN 388:2016, EN 511:2006
Construction Seamless Knit
Coating Material Nitrile
Color Blue, Black
Material Nylon, Polyester
Size Large
Liner Material Acrylic
Resistant To Cold Resistant, Water Resistant, Abrasion Resistant
Series 477
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.