Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (RS)-3,5-DHPG
GREENER_CHOICESDP

Catalog No. 03421R Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-421-R 1 mg
03-421-0 10 mg
03-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 03-421-R Supplier Tocris Bioscience™ Supplier No. 0342/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective group I mGlu agonist

(RS)-3,5-DHPG is a selective group I metabotropic glutamate receptor agonist which activates both mGlu1 and mGlu5. Also reported to be an antagonist at metabotropic glutamate receptors linked to phospholipase D.

S-enantiomer also available.

Specifications

Chemical Name or Material (RS)-3,5-Dihydroxyphenylglycine
CAS 19641-83-9
Assay Percent Range >99%
Quantity 1 mg
Solubility Soluble to 10mM in water
Molecular Formula C8H9NO4
Formula Weight Observed MW: 183.16

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.