Learn More
Honeywell™ Miller™ AirCore™ Steel Harnesses
Catalog No. 19066324 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect
Tongue
D-Ring Location:
Back
None
Side
Includes:
Back D-rings, tongue buckle leg, Quick-connect chest
Quick-Connect chest and leg
Side D-rings, Lumbar Pad and Removable Belt Quick-Connect Chest; Tongue Buckle Leg
Side D-rings, Lumbar Pad and Removable Belt, Quick-Connect Chest and Leg
Side D-rings, quick-connect chest and leg
Side D-rings, tongue buckle leg, quick-connect chest
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) D-Ring Location Includes
19-066-324 Quick-Connect Side Side D-rings, Lumbar Pad and Removable Belt, Quick-Connect Chest and Leg
19-053-137 Quick-Connect Side Side D-rings, quick-connect chest and leg
19-053-138 Tongue Back Back D-rings, tongue buckle leg, Quick-connect chest
19-053-139 Tongue Side Side D-rings, tongue buckle leg, quick-connect chest
19-053-136 Quick-Connect None Quick-Connect chest and leg
19-066-334 Quick-Connect Side Side D-rings, Lumbar Pad and Removable Belt Quick-Connect Chest; Tongue Buckle Leg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-066-324 Supplier Honeywell™ Supplier No. ACQCBDP/UGN
Add to Cart
Add to Cart
Comes in a high-visibility color for greater safety.
  • Lightweight, unique breathable, open-core padding technology provides optimal airflow and reduces heat and moisture build-up up
  • Contoured ergonomic padding design minimizes contact around a worker's neck, lower back and inner thighs to reduce chafing and irritation
  • Patented DuraFlex stretchable webbing provides greater flexibility, increased comfort
  • Cam buckles provide easy one-hand adjustment
  • Holds webbing firmly in place with no readjusting required
  • Stand-up back D-ring easily adjusts and attaches
  • Quick-connect buckles for easy donning
  • Unique and vibrant patterned webbing for easy identification
  • Available with quick connect (QC) or tongue buckle, side-D rings, back-D rings
  • Recommended Industries/Use: Oil and Gas
  • Hazards: Confined Space, fall from height
Certifications
OSHA, ANSI
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI A10.32 and ANSI Z359.1-2007; CSA Z259.10-12 requirements.
Specifications
D-Ring Location Side
Buckle Style (Leg) Quick-Connect
Includes Side D-rings, Lumbar Pad and Removable Belt, Quick-Connect Chest and Leg
Buckle Style (Chest) Quick-Connect