Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Glow-In-The-Dark Electrical Hazard Signs: DANGER - HIGH VOLTAGE

Catalog No. p-4932792 Shop All Brady Corporation Products
Click to view available options
Dimensions (L x W):
25.4 x 17.7 cm
35.5 x 25.4cm
Material:
Plastic
Polyester

Catalog No. 19-000-436

Supplier: Brady™ 80269

Photoluminescent sign to identify electrical hazard and high voltage areas

  • Available in Self-Stick Polyester and Plastic materials
  • Legend: HIGH VOLTAGE

Specifications

Material Plastic
Quantity 1 Ea.
Description Glow-In-The-Dark Plastic
Height (English) 7 in.
Height (Metric) 17.7 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Languages English
Dimensions (L x W) 25.4 x 17.7 cm
Displayed Text Header/Legend: DANGER/HIGH VOLTAGE
Product Type Electrical Hazard Sign
Color Red on Glow
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.