Learn More
Brady™ ALLWIK™ Pillows
Catalog No. p-4933064 Shop All Brady Corporation Products
Click to view available options
Capacity (English):
14 gal.
15.4 gal.
26 gal.
Packaging Quantity:
16/Cs.
32/Cs.
8/Cs.
ALLWIK™ Pillows are used for absorbing spills and leaking fluids in tight spaces.
  • Absorbs all oil and water-based fluids
  • Specifications
    Capacity (English) 26 gal.
    Material Polypropylene
    Type Absorbent Pillow
    For Use With (Application) Overspray leaks and drips, drums - liquid storage, drains
    Packaging Quantity 16/Cs.
    Dimensions (L x W) 45.75 x 45.72 cm (18 x 18 in.)
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.