Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ 4933434E20Rik Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570030
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
4933434E20Rik is a protein coding gene.
Specifications
4933434E20Rik | |
Polyclonal | |
Unconjugated | |
4933434E20Rik | |
4933434E20Rik; 5730552F22Rik; AI462154; NICE-3; NS5ATP4; RIKEN cDNA 4933434E20 gene; uncharacterized protein C1orf43 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
99650 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8R092 | |
4933434E20Rik | |
synthetic peptide directed towards the middle region of human 4933434E20Rik (aa 28-77). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction