Learn More
Bullard™ EVA PAPR System with Double Bib Tychem SL Hood (Saranex Coated)
SDP
Catalog No. p-4939912 Shop All Bullard Company Products
Click to view available options
Facepiece/Headtop Model:
Double bib hood, quick-attach, no headband
Double bib hood, quick-attach, taped seams
Material (Lens):
Acetate
Oversize acetate
Catalog No. 17-989-611
Supplier: Bullard™ EVA20SICT
Only null left
Add to Cart
Add to Cart
Evolutionary Air (EVA) Powered Air-Purifying Respirator (PAPR) uses a battery-powered fan to draw in ambient air through a filter cartridge then delivers filtered air to the user's hood, helmet, or facepiece via a breathing tube
  • Uniquely designed, streamlined form makes PAPR lightweight, eliminates bulky corners that can dig into the wearer's back, and balances the unit's weight for maximum comfort
  • Easy-to-read fuel gauge displays remaining battery capacity at a glance
  • Battery status can be checked at any time during use
  • Bullard Active Flow Technology automatically responds to a worker's need for more or less air flow
  • Two distinct audible alarms notify wearers of low battery and low air flow
  • Alarms are designed so the sound of the alarm travels through the breathing tube directly to the wearer's ear
  • Powered by a long-lasting 10,000-hour brushless motor
  • Single filter design for easy use
  • Filter locking tab notifies the user that the filter is in place with both a visual indication and audible click
Order Info
A NIOSH approved EVA PAPR system consists of the following properly-assembled components: (1) hood or mask; (2) breathing tube; (3) blower assembly; (4) purifying filter cartridge. Check model system components carefully before ordering.
Compliance
NIOSH
Specifications
Battery Life 7 to 8 hours
Facepiece/Headtop Model Double bib hood, quick-attach, taped seams
Includes PAPR system, EVA blower, battery, charger, breathing tube
Material (Lens) Acetate
Model EVA20SICT
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.