Learn More
Corning™ rLaminin-521 (Human)
Catalog No. p-4952224
Click to view available options
Quantity:
100 μg
Recombinant heterotrimer protein composed of α5, β2, and γ1 chains
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
TRUSTED_SUSTAINABILITY
Specifications
Name rLaminin-521, Human
Quantity 100 μg
Source Human
Recombinant Recombinant
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.