Learn More
Abbott™ iScreen™ External Drug Controls
Used as quality control urine to monitor precision of laboratory urine toxicology testing procedures for different analytes
Specifications
Intended Use | Used as quality control urine to monitor precision of laboratory urine toxicology testing procedures |
---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity & Availability | ||||
22-911-083
|
Abbott™
19000102 |
Drug Control Cotinine Specific 2X Positive | 5 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||||
Description
Control designed to work with any urine drug screens
Specifications
Used as quality control urine to monitor precision of laboratory urine toxicology testing procedures |