Learn More
Abbott™ Integrated E-Z Split Key™ Cup A.D. with Built-in Validity Drug Screening
Catalog No. p-4953999
Click to view available options
For Use With (Application):
Detects 10 drugs in urine
Detects 5 drugs in urine
This item is not returnable. View return policy
This item is not returnable. View return policy
Cup with key activated plunger allows to run device as per convenience
  • Integrated cup screens for testing prescription and illicit drugs in urine
  • Integrated specimen validity combinations available: CR, GL, NI, OX, PH, SG
  • Reserved split-specimen ideal for additional confirmation testing
  • Rapid test results in 5 minutes
Specifications
Detectable Analytes AMP, Cocaine, OPI, PCP, THC
Type Integrated E-Z Split Key™ Cup A.D. with Built-in Validity Drug Screening
For Use With (Application) Detects 5 drugs in urine
Quantity 25/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.