Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ BRAND™ BRANDplates™ immunoGrade™ Strip Plates
Click to view available options
Total Volume (Metric) Well:
350 μL
360 μL
Breakapart:
No
Yes
Description
- High-binding, non-sterile plates
- Constructed from high-quality, virgin polystyrene
- Transparent with a flat bottom
- Free from endotoxins, DNase, DNA, RNase; non-cytotoxic
- Available with or without a grid
With grid (Part No. 782306):
- 12 strips of 8 wells each
- Individual strips can be removed from the plate and reinserted
- Strips can also be divided into 8 individual vessels and inserted into the frame plate
Without grid (Part No. 782305):
- 12 strips of 8 wells each
- Individual strips can be removed from the frame and reinserted
- Strips are not divisible
Applications:
Designed for solid phase immunoassays, ELISA, RIA, FIA and other immune testing applications

Specifications
Specifications
Barcode | No Barcode |
Breakapart | No |
Binding Property | immunoGrade |
Certified | DNA, DNase, RNase, Pyrogen free |
Format | Strip Plate |
Array | 1 x 8 |
Autoclavable | Not Autoclavable |
Packaging | Bagged |
Binding Type | High Binding |
Color | Clear |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction