Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Shop All Hamilton Company ProductsDescription
- ideal for dispensing volumes from 0.05 µL up to 5 µL
- The 7000 series is the only zero dead volume syringe
- The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Specifications
| Needle Gauge | 22 G |
| Model | 750 |
| Volume (Metric) | 500 μL |