Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ENV™ Marine Sorbent Booms

Catalog No. p-4955291 Shop All Brady Corporation Products
Click to view available options
Capacity (English):
32 gal./Bale
65 gal./Bale
Dimensions (L x W):
10 ft. x 5 in.
10 ft. x 8 in.

Catalog No. 17-111-089

Supplier: Brady™ ENV510

Designed to float on water and contain or absorb wide variety of marine fuels and oils

  • Feature lint-free blue sleeve
  • Class: Oil only
  • Available in two sizes
  • Specifications

    Capacity (English) 32 gal./Bale
    Material Polypropylene
    Type Sorbent Boom
    Packaging Quantity 4/Cs.
    Dimensions (L x W) 10 ft. x 5 in.
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.