Learn More
Brady™ Oil Only Drag Nets
Catalog No. p-4955348 Shop All Brady Corporation Products
Click to view available options
Includes:
30 nets on 50 ft. rope
30 nets, no rope
Catalog No. 17-111-065
Supplier: Brady™ ODN08
Floats in water and entraps heavy oil
  • Made up of flat, chemical-resistant filaments in half-pound units
  • Can be re-used after draining or dousing
  • Extracted oil can be recycled or downgraded as fuel oil
  • Used as sweep between two boats or deployed along shoreline
  • Available in two sizes: 30 nets on 50 ft. rope and 30 nets (no rope)
Specifications
Type Oil/Drag Net
Includes 30 nets on 50 ft. rope
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.