Learn More
Brady™ Cansorb™ Oil Only Granular Sorbents
Catalog No. p-4955361
Click to view available options
Capacity (English):
10 gal.
20 gal.
Type:
Compressed Granular Sorbent
Loose Granular Sorbent
Ideal for oil-only clean up on open water and land applications
  • Made from fiber sphagnum peat moss
  • Class: Oil only
  • Absorbs 8 times its weight
  • Available in compressed and loose granular format
  • Specifications
    Capacity (English) 20 gal.
    Material Fiber Sphagnum Peat Moss
    Type Compressed Granular Sorbent
    Size 18
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.