Learn More
DuPont™ Tychem™ 6000 Hooded Coverall
Catalog No. 19162808 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
Catalog No. 19-162-808
Supplier: DuPont™ TF145TGY4X0006TV
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Widely used by military personnel and first responders for chemical warfare situations
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
TAA compliant
TRUSTED_SUSTAINABILITY
Specifications
Size 4X-Large
Gender Unisex
Material Tychem
Color Gray
Ankle Style Elastic
Closure Type Zipper
Seam Type Taped
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.