Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InVitroCare™ Sperm Wash Media

Catalog No. p-4955793
Encompass
Click to view available options
Quantity:
100 mL
12 x 12 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For in vitro sperm washing procedures

  • HEPES buffered formulation with 0.5% Human Serum Albumin
  • Does not require use of CO2 incubator
  • Shelf life: 18 months

Specifications

Content And Storage 2°-8°C
Product Type Sperm Wash Medium
With Additives HEPES
Quantity 100 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.