Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ BackBiter Tie-Back Lanyards

Features all-in-one lanyard with SofStop Shock Absorber and cross-arm anchorage connector

Supplier:  Honeywell™ 980B/6FTGN

 View more versions of this product

Catalog No. 17-990-509


Description

Description

  • Innovative BackBiter tie-back lanyard with a 5K Snap Hook
  • Designed from top to bottom for tie-back use
  • Has locking snap and 5K snap hook
  • Contains Double SofStop pack for 3.7m (12 ft.) free fall/duty-rated 400 lb. capacity for up to 1.8m (6 ft.) free fall
  • Available in 6 ft. single and double leg
  • Recommended industries/use: Construction, manufacturing, steel and metals, transportation
  • Hazards: Confined space, fall from height

Certifications

OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6

TRUSTED_SUSTAINABILITY
Specifications

Specifications

400 lb.
Double-Leg
OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6
181.4 kg
1 locking snap hook, 2 5K snap hooks
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.