Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Miller™ BackBiter Tie-Back Lanyards
Features all-in-one lanyard with SofStop Shock Absorber and cross-arm anchorage connector
Specifications
Certifications/Compliance | OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6 |
---|---|
Load Bearing Capacity (English) | 400 lb. |
Load Bearing Capacity (Metric) | 181.4 kg |
Catalog Number | Mfr. No. | Leg Configuration | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Leg Configuration | Includes | Price | Quantity & Availability | ||||
17-990-510
|
Honeywell™
940B/6FTGN |
Single-leg | 1 locking snap hook, 1 5K snap hooks | N/A | N/A | N/A | |||
17-990-509
|
Honeywell™
980B/6FTGN |
Double-Leg | 1 locking snap hook, 2 5K snap hooks | N/A | N/A | N/A | |||
Description
- Innovative BackBiter tie-back lanyard with a 5K Snap Hook
- Designed from top to bottom for tie-back use
- Has locking snap and 5K snap hook
- Contains Double SofStop pack for 3.7m (12 ft.) free fall/duty-rated 400 lb. capacity for up to 1.8m (6 ft.) free fall
- Available in 6 ft. single and double leg
- Recommended industries/use: Construction, manufacturing, steel and metals, transportation
- Hazards: Confined space, fall from height
Certifications
OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6
Specifications
OSHA - 1910.66, 1926.104, 1926.502, ANSI - A10.32, Z359.13, CSA - Z259.11 Class E4, Z259.11 Class E6 | |
400 lb. | |
181.4 kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title