Learn More
Justrite™ Explosion-Proof Heater/Air-Conditioner Package
Shop All Justrite Safety Group Products
Click to view available options
For Use With (Equipment):
12-,16-drum fire rated Agri-Turf lockers
6-,9-drum fire rated Agri-Turf lockers
Description
- For use with 6-,9-,12-,16-drum fire-rated Agri-Turf™ lockers only
- Available in 1.7kWh (6000 BTU) and 3.51kWh (12000 BTU) heater
- 4.4kWh (15000 BTU) and 5.9kWh (20000 BTU) air conditioner
Applications:
Adds heating/air conditioning where comfort, chemical stability or freeze protection is important
†For optimum temperature control, appropriate size R-11 insulation package must be ordered when unit is purchased in conjunction with fire-rated models. This accessory must be purchased in conjunction with locker to ensure proper installation and to retain FM-approval.
Specifications
Specifications
| Product Type | Heater/Air Conditioner Package |
| For Use With (Equipment) | 6-,9-drum fire rated Agri-Turf lockers |