Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter: Probes
Shop All Boekel Scientific Products

Click to view available options
Type:
Conductivity Probe (type k=1.0)
DO Probe
Temperature Probe
pH Probe
Description
For use with Open Source Bio (OSB) Transmitter.
Specifications
Specifications
Type | Temperature Probe |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction