Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4E-BP3 Monoclonal Antibody (1E3)
4E-BP3 Monoclonal Antibody (1E3)
Supplier: Thermo Scientific MA520460
Description
4E-BP3 Monoclonal Antibody for Western Blot, ICC/IF, ELISA

Specifications
4E-BP3 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
O60516 | |
EIF4EBP3 | |
EIF4EBP3 (AAH10881, 1 a.a. ~ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot, Immunocytochemistry | |
1E3 | |
Unconjugated | |
EIF4EBP3 | |
4EBP3; 4E-BP3; eIF4E-binding protein 3; EIF4EBP3; eukaryotic initiation factor 4E-binding protein 3; eukaryotic translation initiation factor 4E binding protein 3; eukaryotic translation initiation factor 4E-binding protein 3; NG36 | |
Mouse | |
Affinity chromatography | |
RUO | |
8637 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction