Learn More
4E-T/eIF4E-T Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300706A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predomitly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| 4E-T/eIF4E-T | |
| Polyclonal | |
| Unconjugated | |
| EIF4ENIF1 | |
| EIF4ENIF1; 4E-T; Clast4; FLJ21601; 2610509L04Rik; eukaryotic translation initiation factor 4E nuclear import factor 1; eIF4E-transporter; FLJ26551; OTTHUMP00000063276 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56478, 74203 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9EST3, Q9NRA8 | |
| Eif4enif1 | |
| Between 925 and C-term | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.