Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4EBP1 Antibody (0014) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$140.50 - $393.00
Specifications
Antigen | 4EBP1 |
---|---|
Clone | 0014 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
4EBP1 Monoclonal antibody specifically detects 4EBP1 in Human samples. It is validated for Western BlotSpecifications
4EBP1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
4E-BP1, eIF4E-binding protein 1, eukaryotic translation initiation factor 4E binding protein 1,4EBP1,4E-BP1BP-1, eukaryotic translation initiation factor 4E-binding protein 1, PHAS-IMGC4316, Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to residues around (Thr37, 46) of Human phospho-4EBP1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0014 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
1978 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title