Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4EBP1 Antibody (2D1G11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$541.95
Specifications
| Antigen | 4EBP1 |
|---|---|
| Clone | 2D1G11 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61901
![]() |
Novus Biologicals
NBP261901 |
0.1 mL |
Each of 1 for $541.95
|
|
|||||
Description
4EBP1 Monoclonal specifically detects 4EBP1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| 4EBP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
| 1978 | |
| Synthesized peptide of human Phospho-4E-BP1 (Ser65). | |
| Primary | |
| 12.6 kDa |
| 2D1G11 | |
| Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| 4E-BP1, eIF4E-binding protein 1, eukaryotic translation initiation factor 4E binding protein 1,4EBP1,4E-BP1BP-1, eukaryotic translation initiation factor 4E-binding protein 1, PHAS-IMGC4316, Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| EIF4EBP1 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title