Learn More
Invitrogen™ 4EBP1 Monoclonal Antibody (4B6G10)

Description
MA5-15313 targets 4E-BP1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15313 immunogen is purified recombinant fragment of 4E-BP1 expressed in E. Coli.
Specifications
Specifications
| Antigen | 4EBP1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 4B6G10 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | EIF4EBP1 |
| Gene Accession No. | Q13541 |
| Gene Alias | 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.