Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98+%, Thermo Scientific Chemicals

Catalog No. p-7046479
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95530-58-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00075172
InChI Key YBUPWRYTXGAWJX-YFKPBYRVSA-N
Synonym r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
PubChem CID 641505
IUPAC Name (4R)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)[C@@H]1COC(=O)N1

Specifications

Melting Point 70°C to 74°C
Beilstein 5376658
Quantity 1 g
Optical Rotation +17° (c=6 in Ethanol)
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material (4R)-(+)-4-Isopropyl-2-oxazolidinone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.