Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone 98.0+%, TCI America™
SDP

Catalog No. p-7131584
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 191090-32-1
Molecular Formula C18H19NO2
Molecular Weight (g/mol) 281.355
MDL Number MFCD03093555
InChI Key PHTOJBANGYSTOH-MRXNPFEDSA-N
PubChem CID 11300512
IUPAC Name (4R)-5,5-diphenyl-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)C1C(OC(=O)N1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 254°C
Color White
Quantity 1 g
Formula Weight 281.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4R)-(+)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.