Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis(4-(trimethylammonio)-phenyl)-21H,23H-porphine tetratosylate, 95%

Catalog No. AC318810250
Click to view available options
Quantity:
25 mg
Packaging:
Glass Bottle

CAS: 69458-20-4 | C84H90N8O12S4 | 1531.928 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69458-20-4
Molecular Formula C84H90N8O12S4
Molecular Weight (g/mol) 1531.928
MDL Number MFCD00012023
InChI Key AKFPMEOALZKHNR-UHFFFAOYSA-J
Synonym ttmapp, 5,10,15,20-tetrakis 4-trimethylammoniophenyl porphyrin tetra p-toluenesulfonate, 5,10,15,20-tetrakis 4-trimethylammonio phenyl-21h,23h-porphine tetra-p-tosylate salt, tetrakis 4-n-trimethylaminophenyl porphine, tetra p-toluenesulfonate ttmapp, purple powder, n,n,n-trimethyl-4-7,12,17-tris 4-trimethylammonio phenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaen-2-yl anilinium tetratosylate
PubChem CID 11979964
IUPAC Name 4-methylbenzenesulfonate;trimethyl-[4-[10,15,20-tris[4-(trimethylazaniumyl)phenyl]-21,22-dihydroporphyrin-5-yl]phenyl]azanium
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+](C)(C)C1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)[N+](C)(C)C)C8=CC=C(C=C8)[N+](C)(C)C)C9=CC=C(C=C9)[N+]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Purple
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 25 mg
Formula Weight 1531.96
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material 5,10,15,20-Tetrakis(4-(trimethylammonio)-phenyl)-21H,23H-porphine tetratosylate, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.