Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Bromoethyl)-2,3-dihydrobenzofuran 98.0+%, TCI America™
SDP

Catalog No. B406625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B406625G 25 g
B40665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B406625G Supplier TCI America Supplier No. B406625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 127264-14-6
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.10
MDL Number MFCD06797641
InChI Key JRKZQRRYNCMSCB-UHFFFAOYSA-N
Synonym 5-(2-Bromoethyl)coumaran
PubChem CID 21831160
IUPAC Name 5-(2-bromoethyl)-2,3-dihydro-1-benzofuran
SMILES BrCCC1=CC2=C(OCC2)C=C1

Specifications

Melting Point 67°C
Quantity 25 g
Formula Weight 227.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Bromoethyl)-2,3-dihydrobenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.