Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(2-Methylphenyl)-1H-tetrazole, 99%

Catalog No. AAL1926203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1926203 1 g
AAL19262MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1926203 Supplier Thermo Scientific Chemicals Supplier No. L1926203
Add to Cart

CAS: 51449-86-6 | C8H8N4 | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51449-86-6
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD03093095
InChI Key MTBUOESFHRORQT-UHFFFAOYSA-N
Synonym 5-2-methylphenyl-1h-tetrazole, 5-o-tolyl-1h-tetrazole, 5-o-tolyl-2h-tetrazole, 5-2-methylphenyl-2h-tetrazole, 5-2-methylphenyl tetrazole, 5-2-methylphenyl-2h-1,2,3,4-tetrazole, 2h-tetrazole,5-2-methylphenyl, 5-2-methylphenyl-1h-1,2,3,4-tetrazole, 2-methylphenyltetrazole, 5-o-tolyl tetrazole
PubChem CID 583067
IUPAC Name 5-(2-methylphenyl)-2H-tetrazole
SMILES CC1=CC=CC=C1C2=NNN=N2

Specifications

Melting Point 151°C to 154°C
Quantity 1 g
Beilstein 136151
Formula Weight 160.18
Percent Purity 99%
Chemical Name or Material 5-(2-Methylphenyl)-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.