Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-[2-(Methylthio)ethyl]hydantoin 99.0+%, TCI America™
SDP

Catalog No. m10461g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 13253-44-6
Molecular Formula C6H10N2O2S
Molecular Weight (g/mol) 174.218
MDL Number MFCD00143392
InChI Key SBKRXUMXMKBCLD-UHFFFAOYSA-N
PubChem CID 567982
IUPAC Name 5-(2-methylsulfanylethyl)imidazolidine-2,4-dione
SMILES CSCCC1C(=O)NC(=O)N1

Specifications

Melting Point 108°C
Color White
Quantity 1 g
Formula Weight 174.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-[2-(Methylthio)ethyl]hydantoin
RTECSNumber NI9461351
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.