Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(3-Pyridyl)-1H-tetrazole, 98%

Catalog No. AAB2049206
Click to view available options
Quantity:
5 g

CAS: 3250-74-6 | C6H5N5 | 147.141 g/mol

5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3250-74-6
Molecular Formula C6H5N5
Molecular Weight (g/mol) 147.141
MDL Number MFCD00040281
InChI Key SECHDFHDDVELCV-UHFFFAOYSA-N
Synonym 3-2h-tetrazol-5-yl pyridine, 5-3-pyridyl-1h-tetrazole, 3-1h-tetrazol-5-yl pyridine, 5-3-pyridyl tetrazole, 3-2h-tetrazol-5-yl-pyridine, pyridine, 3-2h-tetrazol-5-yl, unii-zc84pps834, 3-2h-1,2,3,4-tetrazol-5-yl pyridine, 3-1h-1,2,3,4-tetrazol-5-yl pyridine, 5-3-pyridyl-2h-1,2,3,4-tetraazole
PubChem CID 151091
IUPAC Name 3-(2H-tetrazol-5-yl)pyridine
SMILES C1=CC(=CN=C1)C2=NNN=N2

Specifications

Melting Point ∼240°C (decomposition)
Odor Odorless
Quantity 5 g
Solubility Information Soluble in water (slightly).
Formula Weight 147.14
Percent Purity 98%
Chemical Name or Material 5-(3-Pyridyl)-1H-tetrazole
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
EINECSNumber 426-810-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.