Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Chlorophenyl)-2-furoic acid, 95+%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
41019-45-8
C11H7ClO3
222.63
XIPQHWUSDHTXOO-UHFFFAOYSA-N
5-4-chlorophenyl-2-furoic acid,5-4-chlorophenyl furan-2-carboxylic acid,2-furancarboxylic acid, 5-4-chlorophenyl,5-4-chloro-phenyl-furan-2-carboxylic acid,2-furancarboxylic acid,5-4-chlorophenyl,acid, 62,acmc-1an70,5-p-chlorophenyl furoic acid,5-4-chlorophenyl furoic acid,5-p-chlorophenyl-2furoic acid
726693
5-(4-chlorophenyl)furan-2-carboxylic acid
C1=CC(=CC=C1C2=CC=C(O2)C(=O)O)Cl
41019-45-8
C11H7ClO3
222.63
XIPQHWUSDHTXOO-UHFFFAOYSA-N
5-4-chlorophenyl-2-furoic acid,5-4-chlorophenyl furan-2-carboxylic acid,2-furancarboxylic acid, 5-4-chlorophenyl,5-4-chloro-phenyl-furan-2-carboxylic acid,2-furancarboxylic acid,5-4-chlorophenyl,acid, 62,acmc-1an70,5-p-chlorophenyl furoic acid,5-4-chlorophenyl furoic acid,5-p-chlorophenyl-2furoic acid
726693
5-(4-chlorophenyl)furan-2-carboxylic acid
C1=CC(=CC=C1C2=CC=C(O2)C(=O)O)Cl
Melting Point 198.0°C to 201.0°C
Color Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 95+%
Solubility Information Solubility in water: soluble. Other solubilities: ethanol,acetone,dmf,dioxane
Formula Weight 222.63
Physical Form Powder
Percent Purity 95+%
Chemical Name or Material 5-(4-Chlorophenyl)-2-furoic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning