Learn More
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, 97%
Description
5,5',6,6'-Tetrahydroxy-3,3,3',3'- tetramethyl-1,1'-spirobisindane is used in the preparation of 4,7,4',7'-tetrabromo-3,3,3',3'-tetramethyl-2,3,2',3'-tetrahydro-[1,1']spirobiindene-5,6,5',6'-tetraol by bromination using bromine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5,5′,6,6′-Tetrahydroxy-3,3,3′,3′- tetramethyl-1,1′-spirobisindane is used in the preparation of 4,7,4′,7′-tetrabromo-3,3,3′,3′-tetramethyl-2,3,2′,3′-tetrahydro-[1,1′]spirobiindene-5,6,5′,6′-tetraol by bromination using bromine.
Solubility
Slightly soluble in methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 50 g |
| Beilstein | 2226231 |
| Solubility Information | Slightly soluble in methanol. |
| Formula Weight | 340.42 |
| Percent Purity | 97% |
| Chemical Name or Material | 5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 201-003-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.