Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Diallyl-2,2'-dihydroxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. D39711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39711G 1 g
D3971200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39711G Supplier TCI America Supplier No. D39711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 528-43-8
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00016658
InChI Key VVOAZFWZEDHOOU-UHFFFAOYSA-N
Synonym Magnolol, 5,5′C-Diallylbiphenyl-2,2′C-diol
PubChem CID 72300
ChEBI CHEBI:6643
IUPAC Name 2-(2-hydroxy-5-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES C=CCC1=CC(=C(C=C1)O)C2=C(C=CC(=C2)CC=C)O

Specifications

Melting Point 101°C
Color White
Boiling Point 180°C
Quantity 1 g
Formula Weight 266.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Diallyl-2,2′-dihydroxybiphenyl
RTECSNumber DV5105500
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.