Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethyl-1,3-cyclohexanedione 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
126-81-8
C8H12O2
140.18
MFCD00001588
BADXJIPKFRBFOT-UHFFFAOYSA-N
dimedone,5,5-dimethyl-1,3-cyclohexanedione,cyclomethone,medon,methone,dimedon,methon,1,3-cyclohexanedione, 5,5-dimethyl,5,5-dimethyldihydroresorcinol,5,5-dimethylhydroresorcinol
31358
5,5-dimethylcyclohexane-1,3-dione
CC1(CC(=O)CC(=O)C1)C
126-81-8
C8H12O2
140.18
MFCD00001588
BADXJIPKFRBFOT-UHFFFAOYSA-N
dimedone,5,5-dimethyl-1,3-cyclohexanedione,cyclomethone,medon,methone,dimedon,methon,1,3-cyclohexanedione, 5,5-dimethyl,5,5-dimethyldihydroresorcinol,5,5-dimethylhydroresorcinol
31358
5,5-dimethylcyclohexane-1,3-dione
CC1(CC(=O)CC(=O)C1)C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 500g
Melting Point 147.0°C to 150.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H6(=O)2
Beilstein 07, 559
Fieser 01,266
Merck Index 15, 3264
Solubility Information Solubility in water: 5 % (25°C). Other solubilities: soluble in methanol,ethanol,benzene,chloroform,and acetic acid
Formula Weight 140.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 5, 5-Dimethyl-1, 3-cyclohexanedione, 99%
Show More Show Less

EINECSNumber : 204-804-4

RTECSNumber : GV0345000

TSCA : TSCA