Learn More
5,5'-Dimethyl-2,2'-bipyridine, 98%, Thermo Scientific Chemicals
Description
5,5'-Dimethyl-2,2'-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5’vinyl-2,2’-bipyridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5,5′-Dimethyl-2,2′-bipyridine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is also used as a raw material in the synthesis of dyestuffs and agrochemicals. It is used as a precursor for the preparation of 5-methyl-5’vinyl-2,2’-bipyridine.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 114°C to 117°C |
Boiling Point | 140°C (3 mmHg) |
Quantity | 25 g |
Formula Weight | 184.24 |
Percent Purity | 98% |
Chemical Name or Material | 5,5'-Dimethyl-2,2'-bipyridine |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.