Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane 98.0+%, TCI America™
SDP

Catalog No. D419525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D419525G 25 g
D41955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D419525G Supplier TCI America Supplier No. D419525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5123-13-7
Molecular Formula C11H15BO2
Molecular Weight (g/mol) 190.05
MDL Number MFCD05663851
InChI Key FQMSNYZDFMWLGC-UHFFFAOYSA-N
Synonym (5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzene, Phenylboronic Acid Neopentyl Glycol Ester
PubChem CID 11788741
IUPAC Name 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane
SMILES CC1(C)COB(OC1)C1=CC=CC=C1

Specifications

Melting Point 65°C
Color White
Quantity 25 g
Formula Weight 190.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.