Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Dithiobis-(2-nitrobenzoic acid), 99%

Catalog No. AC117540050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle

CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-78-3
Molecular Formula C14H8N2O8S2
Molecular Weight (g/mol) 396.34
MDL Number MFCD00007140
InChI Key KIUMMUBSPKGMOY-UHFFFAOYSA-N
Synonym dtnb, 5,5'-dithiobis 2-nitrobenzoic acid, ellman's reagent, 3-carboxy-4-nitrophenyl disulfide, dithionitrobenzoic acid, dithiobisnitrobenzoic acid, 5,5'-disulfanediylbis 2-nitrobenzoic acid, benzoic acid, 3,3'-dithiobis 6-nitro, 3,3'-dithiobis 6-nitrobenzoic acid, 5,5'-dithio-bis 2-nitrobenzoic acid
PubChem CID 6254
ChEBI CHEBI:86228
IUPAC Name 5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
SMILES OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O

Specifications

Melting Point 240.0°C to 245.0°C
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula [-SC6H3(NO2)CO2H]2
Quantity 5 g
Beilstein 10, IV, 344
Fieser 01,351
Merck Index 15, 3606
Solubility Information Solubility in water: freely soluble
Formula Weight 396.34
Percent Purity 99%
Chemical Name or Material 5, 5'-Dithiobis-(2-nitrobenzoic Acid)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 200-714-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.