Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Methylenedisalicylic acid, 95%

Catalog No. AC351730250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 122-25-8 | C15H12O6 | 288.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-25-8
Molecular Formula C15H12O6
Molecular Weight (g/mol) 288.26
MDL Number MFCD00016506
InChI Key JWQFKVGACKJIAV-UHFFFAOYSA-N
Synonym 5,5'-methylenedisalicylic acid, 5,5'-methylenebis 2-hydroxybenzoic acid, methylenebis salicylic acid, 5,5-methylenebis salicylic acid, unii-2kf4fvv76n, 5,5-methylenedisalicylic acid, 5-3-carboxy-4-hydroxybenzyl salicylic acid, 3,3'-dicarboxy-4,4'-dihydroxydiphenylmethane, 4,4'-dihydroxy-3,3'-dicarboxydiphenylmethane, 2kf4fvv76n
PubChem CID 67145
IUPAC Name 5-[(3-carboxy-4-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1CC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 235°C to 242°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH2[C6H3(OH)COOH]2
Quantity 25 g
Merck Index 15, 6136
Solubility Information Solubility in water: insoluble in water
Formula Weight 288.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5, 5'-Methylenedisalicylic acid, 95%
Show More Show Less
EINECSNumber 204-530-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.