Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Thiodisalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. T020825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T020825G 25 g
T02085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T020825G Supplier TCI America Supplier No. T020825G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1820-99-1
Molecular Formula C14H10O6S
Molecular Weight (g/mol) 306.288
MDL Number MFCD00020264
InChI Key DWMJRSPNFCPIQN-UHFFFAOYSA-N
PubChem CID 74562
IUPAC Name 5-(3-carboxy-4-hydroxyphenyl)sulfanyl-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1SC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 306.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Thiodisalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.