Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydroquinoline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
10500-57-9
C9H11N
133.19
YQDGQEKUTLYWJU-UHFFFAOYSA-N
2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
66335
5,6,7,8-tetrahydroquinoline
C1CCC2=C(C1)C=CC=N2
10500-57-9
C9H11N
133.19
YQDGQEKUTLYWJU-UHFFFAOYSA-N
2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
66335
5,6,7,8-tetrahydroquinoline
C1CCC2=C(C1)C=CC=N2
Boiling Point 218.0°C to 222.0°C
Quantity 100g
Infrared Spectrum Authentic
Specific Gravity 1.03
Refractive Index 1.544
Flash Point 86°C
Density 1.0300g/mL
Assay Percent Range 97.5% min. (GC)
Beilstein 20,II,176
Formula Weight 133.19
Percent Purity 98%
Chemical Name or Material 5,6,7,8-Tetrahydroquinoline
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 234-030-2