Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6,7,8-Tetrahydroquinoline, 98%

Catalog No. p-7077190
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 10500-57-9 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10500-57-9
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
InChI Key YQDGQEKUTLYWJU-UHFFFAOYSA-N
Synonym 2,3-cyclohexeno pyridine, quinoline, 5,6,7,8-tetrahydro, unii-l786aag56h, 5,6,7,8-tetra-hydroquinoline, 5,6,7,8-tetrahydro-quinoline, pubchem5891, 2,3-cyclohexenopyridine, g00034-watson-int, 2,3-cyclohexano pyridine, acmc-2098eh
PubChem CID 66335
IUPAC Name 5,6,7,8-tetrahydroquinoline
SMILES C1CCC2=C(C1)C=CC=N2

Specifications

Density 1.0300g/mL
Boiling Point 218.0°C to 222.0°C
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.544
Quantity 5 g
Beilstein 20,II,176
Specific Gravity 1.03
Formula Weight 133.19
Percent Purity 98%
Chemical Name or Material 5,6,7,8-Tetrahydroquinoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 234-030-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.